Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse DLL4 ELISA Kit

Catalog No. EM21RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM21RBX10 10 x 96 Tests
EM21RBX5 5 x 96 Tests
EM21RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM21RBX10 Supplier Invitrogen™ Supplier No. EM21RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Mouse DLL4 quantitates mouse DLL4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse DLL4.

Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AOS6; delta 4; delta ligand 4; delta like canonical Notch ligand 4; Delta4; delta-like 4 (Drosophila); delta-like 4 homolog; delta-like 4 protein; delta-like protein 4; DLL4; Drosophila Delta homolog 4; hdelta2; MGC126344; notch ligand delta-2; notch ligand DLL4; UNQ1895/PRO4341
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 54485
Gene Symbol Dll4
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target DLL4
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.