Learn More
ACROBiosystems Mouse DM-1 25ug
Supplier:  ACROBiosystems DM1BLY7325UG
Encompass_Preferred
Mertansine (DM-1) is a tubulin inhibitor that binds to the ends of microtubules and inhibits microtubule dynamics. DM-1(Mertansine) has antitumor activity and functions as a regulator of tubulin. It is an alpha-amino acid ester a carbamate an epoxide an organic heterocyclic tetracyclic compound an organochlorine compound a mercaptan and a maydenin alkaloid. DM-1 derived from Mydenin is a cytotoxic component of antibody-drug conjugations that produce antibody-drug conjugations via a sulfhydryl group splice with SPP (n-succinimide 4- (2-pyridyl dithio)) or SMCC (4- (3-mercapto-2 5-dioxy-1 pyrrolidyl) -cyclohexanic acid) splice.
Catalog No. 50-253-5171
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.