Learn More
R&D Systems™ Mouse Dopamine D3 R/DRD3 (NP_031903.1) VersaClone cDNA

Shop All Novus Biologicals Products


Specifications
Specifications
| Gene | Dopamine D3R/DRD3 |
| Accession Number | NP_031903.1 |
| Gene ID (Entrez) | 1814 |
| Gene Alias | D(3) dopamine receptor, D3DR, Dopamine D3 receptor, Dopamine D3R, dopamine receptor D3, DRD3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.