Learn More
American Research Products Inc Mouse Ectodysplasin A EDA EL
Supplier:  American Research Products Inc CSBEL007389MO
Encompass
Mouse Ectodysplasin-A (EDA) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28050