Description
Feeder layers of mouse embryonic fibroblasts (MEF) support stem cell growth and the maintenance of pluripotency by secreting growth factors, cytokines and nutrients. However, the use of MEFs as a feeder layer can be laborious and MEF contamination is a significant concern. MEF conditioned media represents an efficient alternative to MEF feeder cells.
- Eliminates the need for a MEF feeder layer
- Lot-to-lot consistency decreases experimental variability
- Addition of FGF basic generates complete media
- Free of mycoplasma and microbial contamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Stem Cell Conditioned Media |
| Product Line | R&D Systems |
| Quantity | 100 mL |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |