Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Endothelin-1 ELISA Kit

Catalog No. EM26RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM26RBX10 10 x 96 Tests
EM26RBX5 5 x 96 Tests
EM26RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM26RBX10 Supplier Invitrogen™ Supplier No. EM26RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Mouse Endothelin-1 quantitates mouse Endothelin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Endothelin-1.

The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; Et1; ET-1; HDLCQ7; PPET1; PPET-1; preproendothelin; preproendothelin-1; preproET; QME
Assay Range 4.5 to 1,000 pg/mL
Assay Sensitivity 4.5 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 13614
Gene Symbol EDN1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Endothelin 1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.