Learn More
Sino Biological EphA2 / Eph Receptor A2 Antibody (PE), Rabbit MAb 25Tests
Supplier:  Sino Biological 50586R101P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 / Eph Receptor A2 (rM EphA2 / Eph Receptor A2; Catalog#50586-M08H; NP_034269.2; Met1-Asn535) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-3201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.