Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse EphA3 Fc Chimera Recombinant Protein

Catalog No. PIRP88231
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88231 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88231 Supplier Invitrogen™ Supplier No. RP88231
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (800 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

EPHA3 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA3 encodes a protein that binds ephrin-A ligands and receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29319
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 13837
Molecular Weight (g/mol) 96 kDa
Name Mouse EphA3 Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 200 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias AW492086; Cek4; EK4; End3; EPH receptor A3; EPHA3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; H TYRO4; Hek; Hek4; Human embryo kinase; human embryo kinase 1; I79_002835; Mek4; rEK4; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase TYRO4
Common Name EphA3
Gene Symbol EPHA3
Biological Activity Measured by its binding ability in a functional ELISA. Immobilized mouse EFNA5-His (at 10 μg/ml (100 μL/well) can bind mouse EPHA3-Fc, The EC50 of rat mouse EPHA3-Fc is 12.3-28.9 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Mouse EPHA3, amino acids Met1-His541 (Accession # NP_034270.1) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured by its binding ability in a functional ELISA. Immobilized mouse EFNA5-His at 10 μg/ml (100 μL/well) can bind mouse EPHA3-Fc, The EC50 of rat mouse EPHA3-Fc is 12.3-28.9 ng/mL.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.