Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Erythropoietin/EPO Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067990
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9591SP 25 μg
MAB9591100 100 μg
MAB9591500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB9591SP Supplier R&D Systems Supplier No. MAB9591SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Erythropoietin/EPO
Applications Neutralization, ELISA
Classification Monoclonal
Clone 148436
Conjugate Unconjugated
Dilution Neutralization 0.15-0.75 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P07321
Gene Alias ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Gene Symbols EPO
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Erythropoietin/EPO, Ala27-Arg192, Accession # P07321
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2056
Test Specificity Detects mouse Erythropoietin/EPO in direct ELISAs. In direct ELISAs, approximately 25-50% cross-reactivity with recombinant rat Erythropoietin/EPO, 5-15% cross-reactivity with recombinant human Erythropoietin/EPO, and no cross-reactivity with recombinant mouse Thrombopoietin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.