Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Exostosin-like 1/EXTL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7065985
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4214SP 25 μg
MAB4214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4214SP Supplier R&D Systems Supplier No. MAB4214SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Exostosin-like 1/EXTL1 Monoclonal specifically detects Exostosin-like 1/EXTL1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Exostosin-like 1/EXTL1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 353713
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9JKV7
Gene Alias alpha-N-acetylglucosaminyltransferase II, EC 2.4.1.224, exostoses (multiple)-like 1, Exostosin like 1, Exostosin-L, EXTL1, EXTLexostosin-like 1, glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Glucuronosyl-N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase, glucuronyl-N-acetylglucosaminylproteoglycan alpha-1,4-N-acetylglucosaminyltransferase, MGC70794, Multiple exostosis-like protein
Gene Symbols EXTL1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 1/EXTL1 Ala30-Pro669 Accession # Q9JKV7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2134
Test Specificity Detects mouse Exostosin-like 1/EXTL1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) EXTL1, rhEXTL2, rhEXTL3, or recombinant mouse EXTL2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.