Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Exostosin-like 2/EXTL2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7065144
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2536SP 25 μg
MAB2536 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2536SP Supplier R&D Systems Supplier No. MAB2536SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Exostosin-like 2/EXTL2 Monoclonal specifically detects Exostosin-like 2/EXTL2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Exostosin-like 2/EXTL2
Applications Western Blot
Classification Monoclonal
Clone 339509
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9ES89
Gene Alias Alpha-1,4-N-acetylhexosaminyltransferase EXTL2, alpha-1,4-N-acteylhexosaminyltransferase, Alpha-GalNAcT EXTL2, EC 2.4.1.223, exostoses (multiple)-like 2, Exostosin like 2, exostosin-like 2, EXTL2, EXTR2, EXT-related protein 2, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, processed exostosin-like 2
Gene Symbols EXTL2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 2/EXTL2 Asn43-Met330 Accession # Q9ES89
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2135
Test Specificity Detects mouse Exostosin-like 2/EXTL2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse EXTL1, recombinant human (rh) Exostosin-like 2/EXTL2, or rhEXTL3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.