Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Factor XII ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB030334 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB030334 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB030334 Supplier Novus Biologicals™ Supplier No. NBP282405
Add to Cart
Add to Cart

Mouse Factor XII ELISA Kit (Chemiluminescence) measures Mouse Factor XII in Serum, plasma, cell supernatant, and other biological fluids.

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse Factor XII in samples. No Significant cross-reactivity or interference between Mouse Factor XII and analogues was observed.
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 2161
Gene Symbol F12
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, F12, HAE3, HAEX, HAF, HAF beta-factor XIIa part 2, Hageman factor
Target Factor XII
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.