Learn More
Description
- Fas Fas (TNF receptor superfamily member 6) [ Mus musculus (house mouse) ]
- Also known as: lpr; APO1; APT1; CD95; TNFR6; Tnfrsf6; AI196731
- FAS belongs to the death receptor subfamily of the TNF receptor superfamily.
- It has been shown to play a central role in the physiological regulation of programmed cell death and has been implicated in the pathogenesis of various malignancies and diseases of the immune system.
- The interaction of FAS with its ligand allows the formation of a deathinducing signaling complex that includes Fas-associated death domain protein (FADD), caspase 8, and caspase 10.
- The autoproteolytic processing of the caspases in the complex triggers a downstream caspase cascade and leads to apoptosis.
- The Fas/TNFRSF6/CD95 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mFas |
| Accession Number | NP_032013 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 997 bp |
| Gene ID (Entrez) | 355 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
