Learn More
Sino Biological Mouse FCGR1 MAb 100TESTS 50086-R027-A-100
Supplier:  Sino Biological 50086R027A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD64 / FCGR1 (rM CD64 / FCGR1; Catalog#50086-M08H; NP_034316.1; Met 1-Pro 297) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-9161
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.