Learn More
Sino Biological Mouse FCGRT & B2M Heterodimer Protein, Endotoxin-Free 100µg
Supplier:  Sino Biological CT029M08HUE100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 297) of mouse FCGRT (Q61559) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the mouse B2M (P01887,105Ala/Asp) (Met 1-Met 119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT / B2M heterodimer was purified
Catalog No. 50-000-83613
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.