Learn More
Invitrogen™ Mouse Fetuin A ELISA Kit
Description
Mouse Fetuin A quantitates mouse Fetuin A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Fetuin A.
Specifications
Specifications
| Accession Number | 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743 |
| Assay Range | 8 to 2,000 ng/mL |
| Assay Sensitivity | 8 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 11625 |
| Gene Symbol | AHSG |
| Interassay CV | <12% |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.