Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse FGF-21 Recombinant Protein, PeproTech®

Catalog No. 450561MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-56-1MG 1 mg
45056100UG 100 μg
45056250UG 250 μg
450-56-25UG 25 μg
45056500UG 500 μg
450-56-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 450-56-1MG Supplier Gibco™ Supplier No. 450561MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine FGF-21 is a 20.1 kDa protein containing 182 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9JJN1
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 56636
Molecular Weight (g/mol) 20.1 kDa
Name Mouse FGF-21
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196
Common Name FGF21
Gene Symbol FGF21
Biological Activity Determined by its ability to stimulate the proliferation of murine NIH-3T3 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MAYPIPDSSP LLQFGGQVRQ RYLYTDDDQD TEAHLEIRED GTVVGAAHRS PESLLELKAL KPGVIQILGV KASRFLCQQP DGALYGSPHF DPEACSFREL LLEDGYNVYQ SEAHGLPLRL PQKDSPNQDA TSWGPVRFLP MPGLLHEPQD QAGFLPPEPP DVGSSDPLSM VEPLQGRSPS YAS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.