Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse FGF-23 Recombinant Protein, PeproTech®

Catalog No. 4505501M
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-055-01M 2 x 500 μg
45055100UG 100 μg
450-55-20UG 20 μg
45055250UG 250 μg
45055500UG 500 μg
450-55-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 45-055-01M Supplier Gibco™ Supplier No. 4505501M
Add to Cart
Add to Cart

Recombinant Protein

450-55-1MG will be provided as 2 x 500 μg (450-55-500UG). Recombinant Murine FGF-23 is a 25.5 kDa globular protein containing 228 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9EPC2
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 64654
Molecular Weight (g/mol) 25.5 kDa
Name Mouse FGF-23
Purification Method Purified
Quantity 2 x 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828
Common Name FGF23
Gene Symbol FGF23
Biological Activity Determined by its ability to stimulate the proliferation of murine NIH-3T3 cells. The expected ED50 for this effect is 0.3-0.5 ug/ml, in the presence of murine Klotho and heparin.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MYPDTSPLLG SNWGSLTHLY TATARTSYHL QIHRDGHVDG TPHQTIYSAL MITSEDAGSV VITGAMTRRF LCMDLHGNIF GSLHFSPENC KFRQWTLENG YDVYLSQKHH YLVSLGRAKR IFQPGTNPPP FSQFLARRNE VPLLHFYTVR PRRHTRSAED PPERDPLNVL KPRPRATPVP VSCSRELPSA EEGGPAASDP LGVLRRGRGD ARGGAGGADR CRPFPRFV
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of murine NIH-3T3 cells. The expected ED50 for this effect is 0.3-0.5 ug/ml, in the presence of murine Klotho and heparin.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.