Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse FGF-9 Recombinant Protein

Catalog No. PIRP8637
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8637 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8637 Supplier Invitrogen™ Supplier No. RP8637

Recombinant Protein

Fibroblast Growth Factor 9 (mouse) Recombinant Protein, RP-8637, from Invitrogen.

Fibroblast Growth Factor-9 (FGF-9) is a steroid-regulated mitogen and survival factor for nerve and mesenchymal cells. FGF-9 is an autocrine/paracrine growth factor considered to be important for the growth and survival of motorneurons and prostate tissue. Recombinant mouse G-CSF is a non-glycosylated protein, containing 179 amino acids, with a molecular weight of 19 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent proliferation of mouse M-NFS-60 cells and is typically less than 10-60 ng/mL. The specific activity of this protein is 1x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P54130
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 10mM sodium phosphate with no preservative; pH 7.5
Gene ID (Entrez) 14180
Molecular Weight (g/mol) ∽23.3
Name Mouse FGF9
pH Range 7.5
Quantity 10 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias M-FGF-9
Common Name Mouse FGF9
Gene Symbol Fgf9
Specific Reactivity Mouse
Conjugate Unconjugated
Cross Reactivity Ms
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitroresearch only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.