Learn More
Sino Biological Mouse FGF21 / Fibroblast Growth Factor 21 Protein (His Tag) 1mg
Supplier:  Sino Biological 50421M08H1
Encompass_Preferred
A DNA sequence encoding the mouse FGF21 (Q9JJN1) (Met 1-Ser 210) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3276
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.