Learn More
R&D Systems™ Mouse Fibrinogen Degradation Product ELISA Kit (Chemiluminescence)
Shop All Novus Biologicals Products

Specifications
Specifications
| Assay Range | 31.25 to 2000 ng/mL |
| Assay Sensitivity | 18.75 ng/mL |
| Detection Method | Chemiluminescence |
| Product Line | Novus |
| Sample Type | Serum, Plasma, Other Biological Fluids |
| Specificity | This kit recognizes Mouse Fibrinogen Degradation Product in samples. No significant cross-reactivity or interference between Mouse Fibrinogen Degradation Product and analogues was observed |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.