Learn More
Kamiya Biomedical KAMIYA Mouse Fibrinogen ELISA, Highly Sensitive Double Antibody Sandwich Assay, Suitable for Mouse Samples, 96 Tests
SDP
Supplier:  Kamiya Biomedical KT397
Encompass
The KAMIYA BIOMEDICAL COMPANY Mouse Fibrinogen ELISA is a sensitive enzyme-linked immunoassay for quantitatively measuring fibrinogen levels in mouse biological samples. It uniquely employs a double antibody sandwich method, allowing for precise determination of fibrinogen as an acute phase reactant. The colorimetric detection at 450 nm, along with its broad assay range of 25 - 800 ng/mL, makes it an essential tool for researchers in the field.
Catalog No. NC3245756