Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Galectin-3 DuoSet ELISA
SDP

Catalog No. DY119705 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY119705 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY119705 Supplier R&D Systems™ Supplier No. DY119705
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Galectin-3 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Galectin-3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Galectin-3.

Specifications

Assay Range 15.60 to 1000 pg/mL
Product Type Solid Phase Sandwich ELISA
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 3958
Gene Symbol Lgals3
Quantity 1 Kit (for 5 Plates)
Sample Volume 100 μL
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3
Target Galectin-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.