Learn More
Abcam Mouse Galectin 3 ELISA Kit (ab203369)
Supplier:  Abcam AB2033691X96TESTS
Encompass_Preferred
Mouse Galectin 3 ELISA Kit (ab203369) |Key features and details---One-wash 90 minute protocol |Sensitivity: 2.4 pg/ml |Range: 7.81 pg/ml - 500 pg/ml |Sample type: Cell culture supernatant, Cit plasma, EDTA Plasma, Hep Plasma, Serum, Urine |Detection method: Colorimetric |Assay type: Sandwich (quantitative) |Reacts with: Mouse |
Catalog No. NC2102956
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.