Learn More
Rockland Immunochemicals MOUSE Gamma Globulin Fraction - D609-0100
SDP
Supplier:  Rockland Immunochemicals D6090100
Encompass
Gamma globulins are a class of globulins, identified by their position after serum protein electrophoresis. The most significant gamma globulins are immunoglobulins ("Igs"), more commonly known as antibodies, although some Igs are not gamma globulins, and some gamma globulins are not Igs. Globulin is one of the three main types of serum proteins, the others being albumin and fibrinogen. Some globulins are produced in the liver, while others are made by the immune system.
Catalog No. NC9333390