Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse GDF-5 Recombinant Protein, PeproTech®

Catalog No. 31524100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31524100UG 100 μg
315-24-10UG 10 μg
315-24-1MG 1 mg
31524250UG 250 μg
31524500UG 500 μg
315-24-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 31524100UG Supplier Gibco™ Supplier No. 31524100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine GDF-5 is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43027
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 14563
Molecular Weight (g/mol) 27 kDa
Name Mouse GDF-5
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; Cdmp-1; GDF5; GDF-5; growth differentiation factor 5; growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; radotermin; SYM1B; SYNS2
Common Name GDF5
Gene Symbol GDF5
Biological Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 1.0-2.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence APLANRQGKR PSKNLKARCS RKALHVNFKD MGWDDWIIAP LEYEAFHCEG LCEFPLRSHL EPTNHAVIQT LMNSMDPEST PPTCCVPTRL SPISILFIDS ANNVVYKQYE DMVVESCGCR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.