Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GDF-8/Myostatin Propeptide Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF1539SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1539SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1539SP Supplier R&D Systems Supplier No. AF1539SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 7 publications

GDF-8/Myostatin Polyclonal specifically detects GDF-8/Myostatin in Human∣Mouse∣Transgenic Mouse samples. It is validated for Immunoprecipitation∣Neutralization∣Western Blot.

Specifications

Antigen GDF-8/Myostatin
Applications Immunoprecipitation, Neutralization, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O08689
Gene Alias GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Gene Symbols MSTN
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GDF-8/Myostatin Propeptide Asn25-Ser265 Accession # O08689
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2660
Test Specificity Detects GDF-8/Myostatin Propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with mature recombinant mouse (rm) GDF-8 is observed and less than 5% cross-reactivity with mature rmGDF-1, -3, -5, -6, -7, -9, and recombinant human GDF-11 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Transgenic Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.