Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse GDF-8/Myostatin Propeptide Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1539
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1539 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1539 Supplier R&D Systems Supplier No. AF1539
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 7 publications

GDF-8/Myostatin Polyclonal specifically detects GDF-8/Myostatin in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen GDF-8/Myostatin
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL
Gene Accession No. O08689
Gene Alias GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Gene Symbols MSTN
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GDF-8/Myostatin Propeptide Asn25-Ser265 Accession # O08689
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2660
Test Specificity Detects GDF-8/Myostatin Propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with mature recombinant mouse (rm) GDF-8 is observed and less than 5% cross-reactivity with mature rmGDF-1, -3, -5, -6, -7, -9, and recombinant human GDF-11 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.