Learn More
Gibco™ Mouse GDNF, Animal-Free Recombinant Protein, PeproTech®
Description
The functional murine GDNF ligand is a disulfide-linked homodimer consisting of two 15.1 kDa polypeptide chains called monomers. Each monomer contains seven conserved cysteine residues, including Cys-101, which is used for inter-chain disulfide bridging, and others that are involved in the intramolecular ring formation known as the cysteine-knot configuration. The calculated molecular weight of Recombinant Murine GDNF is 30.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P48540 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 14573 |
| Molecular Weight (g/mol) | 30.2 kDa |
| Name | Mouse GDNF, Animal-Free |
| Quantity | 500 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.