Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse GDNF Recombinant Protein, PeproTech®

Catalog No. 4504410UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-44-10UG 10 μg
45044100UG 100 μg
450-44-1MG 1 mg
45044250UG 250 μg
450-44-2UG 2 μg
45044500UG 500 μg
450-44-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 450-44-10UG Supplier Gibco™ Supplier No. 4504410UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

The functional murine GDNF ligand is a disulfide-linked homodimer consisting of two 15.1 kDa polypeptide chains called monomers. Each monomer contains seven conserved cysteine residues, including Cys-101, which is used for inter-chain disulfide bridging, and others that are involved in the intramolecular ring formation known as the cysteine-knot configuration. The calculated molecular weight of Recombinant Murine GDNF is 30.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48540
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 14573
Molecular Weight (g/mol) 30.2 kDa
Name Mouse GDNF
Quantity 10 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor
Common Name GDNF
Gene Symbol GDNF
Biological Activity The ED50 was determined by the proliferation of rat C6 cells is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MSPDKQAAAL PRRERNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.