Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse GGT1 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP349741 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP349741 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP349741 Supplier Novus Biologicals™ Supplier No. NBP349741
Add to Cart
Add to Cart

Colorimetric Assay Kit

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Detection Method Chemiluminescence
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Mouse GGT1 in samples. No significant cross-reactivity or interference between Mouse GGT1 and analogues was observed
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 2678
Gene Symbol GGT1
Quantity 1 Kit
Research Discipline Cancer
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD224, CD224 antigen, D22S672, D22S732, EC 2.3.2.2, gamma-glutamyl transpeptidase, gamma-glutamyltransferase 1 mgC96904, gamma-glutamyltranspeptidase 1, GGT 1, GGTMGC96963, glutamyl transpeptidase, GTG, MGC96892
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.