Learn More
Crystal Chem Inc Mouse Glucagon ELISA Kit, 96-wells, Uses 10uL of sample
Supplier: Crystal Chem Inc 81518
The Mouse Glucagon ELISA is used to quantify levels of mouse glucagon in plasma, serum, and cell culture using only a 10 µL sample volume. Highlights: Kits use a sample volume of only 10 µL. High Sensitivity of 1.1 pg/mL. Dynamic Range of 7.8 - 500 pg/mL. Designed for mouse samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.