Learn More
Description
Includes
Mouse GM-CSF Capture Antibody; Mouse GM-CSF Detection Antibody; Mouse GM-CSF Standard; Streptavidin-HRP
Specifications
Specifications
| Conjugate | Biotin |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Quantity | 5 Plates |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
