Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse gp130 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5380107
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4682SP 25 μg
MAB4682100 100 μg
MAB4682500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB4682SP Supplier R&D Systems Supplier No. MAB4682SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

gp130/CD130 Monoclonal specifically detects gp130/CD130 in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen gp130/CD130
Applications Neutralization, ELISA
Classification Monoclonal
Clone 125605
Conjugate Unconjugated
Dilution Neutralization, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q00560
Gene Alias CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Gene Symbols IL6ST
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse gp130 Gln23-Glu617 Accession # Q00560
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3572
Test Specificity Detects mouse gp130 ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human gp130, recombinant mouse (rm) rmIL-6, or rmIL-6R is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.