Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse GPR15L Recombinant Protein, PeproTech®

Catalog No. 30072100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30072100UG 100 μg
300-72-1MG 1 mg
30072250UG 250 μg
300-72-25UG 25 μg
30072500UG 500 μg
300-72-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30072100UG Supplier Gibco™ Supplier No. 30072100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's E.coli-derived Recombinant Murine GPR15L consists of 54 amino acid residues and has a calculated molecular weight of 6.3 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GPR15L is a newly identified ligand for GPR15, a member of the G protein-coupled receptor (GPCR) family. Upon ligation, GPR15L acts as a potent chemoattractant for GPR15-expressing T cells and together they mediate lymphocyte recruitment to the large intestine and skin. GPR15L is constitutively expressed by colon epithelial cells where its expression is minimally altered by intestinal inflammation. Conversely, GPR15L is nearly undetectable in adult epidermis but highly upregulated during wound healing and inflammation, particularly in psoriasis. Significant expression of GPR15L is also seen in additional mucosal epithelial cells, including those of the stomach, esophagus, and urinary tract. While maintaining similar expression patterns and intramolecular disulfide cysteine bridges found in members of the CC chemokine family, GPR15L differs from classic CC and CXC chemokines, whose active sites are found on the N-terminus, in that it relies on its C-terminus for receptor interaction. GPR15L was first identified as an antimicrobial peptide (AMP) due to its broad spectrum of antimicrobial activity, a property shared with many chemokines. GPR15L binds to an additional receptor, SUSD2, and early studies have indicated that both are downregulated in colon cancer tissue resulting in inhibited colon cancer growth.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation protein with no preservative
Molecular Weight (g/mol) 6.3 kDa
Name Mouse GPR15L
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Antimicrobial peptide with 57 amino acid residues; anti-microbial peptide with 57 amino acid residues; antimicrobial peptide-57; AP-57; C10orf99; chromosome 10 open reading frame 99; colon-derived SUSD2 binding factor; CSBF; GPR15L; Protein GPR15L; RLLV1833; secreted protein C10orf99; UNQ1833; UNQ1833/PRO3446
Common Name C10orf99
Conjugate Unconjugated
Recombinant Recombinant
Sequence RRHPAKSLKL RRCCHLSPRS KLTTWKGNHT RPCRLCRNKL PVKSWVVPGA LPQI
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.