Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Mouse Granzyme B DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Granzyme B.
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3002 |
| Gene Symbol | Gzmb |
| Host Species | Mouse |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT |
| Target | Granzyme B |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
