Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Granzyme G Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064333
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1346SP 25 μg
MAB1346 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1346SP Supplier R&D Systems Supplier No. MAB1346SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Granzyme G Monoclonal specifically detects Granzyme G in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Granzyme G
Applications Western Blot
Classification Monoclonal
Clone 207639
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P13366
Gene Alias Ctla7, Ctla-7, GrzG, GZMG
Gene Symbols GZMG
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Granzyme G Ile21-Leu248 Accession # P13366
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14944
Test Specificity Detects mouse Granzyme G in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse Granzyme D is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.