Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Gremlin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB956SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB956SP 25 μg
MAB956100 100 μg
MAB956500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB956SP Supplier R&D Systems Supplier No. MAB956SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Gremlin 1 Monoclonal specifically detects Gremlin 1 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Gremlin 1
Applications ELISA
Classification Monoclonal
Clone 155606
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O70326
Gene Alias Cell proliferation-inducing gene 2 protein, CKTSF1B1gremlin 1, cysteine knot superfamily, homolog, Cysteine knot superfamily 1, BMP antagonist 1gremlin 1, cysteine knot superfamily, homolog (Xenopus laevis), DAN domain family member 2, DAND2, DAND2GREMLIN, Down-regulated in Mos-transformed cells protein, DRM, DRMMGC126660, GREM1, gremlin, gremlin 1, gremlin 1-like protein, gremlin-1, IHG-2, Increased in high glucose protein 2, increased in high glucose-2, proliferation-inducing gene 2
Gene Symbols GREM1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Gremlin Lys25-Asp184 Accession # O70326
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26585
Test Specificity Detects mouse Gremlin in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant chicken Caronte, recombinant human (rh) Cerberus, rhCOCO, rhGremlin, rhBMP-2, rhBMP-4, recombinant mouse (rm) DAN, rmPRDC, rmSlit1, rmSlit2, or rmBMP-4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.