Learn More
Bioworld MOUSE GRO/KC (CXCL1) (RECO
Supplier:  Bioworld 220605721
Encompass_Preferred
MOUSE GRO/KC (CXCL1), (RECOMBINANT): Recombinant Mouse GRO1/N51/KC (CXCL1) Source: E.Coli Background: KC is a member of the CXC chemokine family with homology to human IL-8, a potent neutrophilic chemotactic cytokine. KC is a fundame...
Catalog No. 50-253-672
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.