Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse H60 ELISA Kit

Catalog No. EM35RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM35RB 96 Tests
EM35RBX5 5 x 96 Tests
EM35RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM35RB Supplier Invitrogen™ Supplier No. EM35RB
Add to Cart
Add to Cart

ELISA

Mouse H60 quantitates mouse H60 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse H60.

Enables natural killer cell lectin-like receptor binding activity. Acts upstream of or within several processes, including positive regulation of immune response; positive regulation of interferon-gamma production; and positive regulation of leukocyte activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number H60; H60a; histocompatibility 60; histocompatibility 60 A
Assay Range 82 to 20 ng/mL
Assay Sensitivity 82 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 15101
Gene Symbol H60a
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target H60
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.