Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Hepcidin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB95051SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB95051-SP 25 μg
MAB95051-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB95051-SP Supplier R&D Systems Supplier No. MAB95051SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Hepcidin Antimicrobial Peptide Monoclonal specifically detects Hepcidin Antimicrobial Peptide in Mouse samples. It is validated for ELISA.

Specifications

Antigen HepcidinAntimicrobialPeptide
Applications ELISA
Classification Monoclonal
Clone 968018
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q9EQ21
Gene Alias HAMP, HEPC, hepcidin antimicrobial peptide, HEPCPutative liver tumor regressor, HFE2B, HFE2Bhepcidin, LEAP1, LEAP-1, LEAP1PLTR, Liver-expressed antimicrobial peptide 1, PLTR
Host Species Rat
Immunogen Synthetic peptide containing mouse Hepcidin Accession # Q9EQ21
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 57817
Test Specificity Detects mouse Hepcidin in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.