Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse HGF Recombinant Protein, PeproTech®

Catalog No. 3152350UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 250 μg
20 μg
250 μg
4 x 250 μg
5 μg
50 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-23-50UG 50 μg
31523100UG 100 μg
31-523-01M 4 x 250 μg
31-523-500 2 x 250 μg
315-23-20UG 20 μg
31523250UG 250 μg
315-23-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 315-23-50UG Supplier Gibco™ Supplier No. 3152350UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

315-23-1MG will be provided as 4 x 250 μg (315-23-250UG). 315-23-500UG will be provided as 2 x 250 μg (315-23-250UG). Recombinant Murine HGF is a 79.3 kDa polypeptide consisting of 696 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08048
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 15234
Molecular Weight (g/mol) 79.3 kDa
Name Mouse HGF
Purification Method Purified
Quantity 50 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF
Common Name HGF
Gene Symbol Hgf
Biological Activity Determined by the dose-dependent stimulation of the proliferation of mouse IMCD3 cells using a concentration range of 10-20 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Alpha chain: QKKRRNTLHE FKKSAKTTLT KEDPLLKIKT KKVNSADECA NRCIRNRGFT FTCKAFVFDK SRKRCYWYPF NSMSSGVKKG FGHEFDLYEN KDYIRNCIIG KGGSYKGTVS ITKSGIKCQP WNSMIPHEHS FLPSSYRGKD LQENYCRNPR GEEGGPWCFT SNPEVRYEVC DIPQCSEVEC MTCNGESYRG PMDHTESGKT CQRWDQQTPH RHKFLPERYP DKGFDDNYCR NPDGKPRPWC YTLDPDTPWE YCAIKTCAHS AVNETDVPME TTECIQGQGE GYRGTSNTIW NGIPCQRWDS QYPHKHDITP ENFKCKDLRE NYCRNPDGAE SPWCFTTDPN IRVGYCSQIP KCDVSSGQDC YRGNGKNYMG NLSKTRSGLT CSMWDKNMED LHRHIFWEPD ASKLNKNYCR NPDDDAHGPW CYTGNPLIPW DYCPISRCEG DTTPTIVNLD HPVISCAKTK QLR Beta chain: VVNGIPTQTT VGWMVSLKYR NKHICGGSLI KESWVLTARQ CFPARNKDLK DYEAWLGIHD VHERGEEKRK QILNISQLVY GPEGSDLVLL KLARPAILDN FVSTIDLPSY GCTIPEKTTC SIYGWGYTGL INADGLLRVA HLYIMGNEKC SQHHQGKVTL NESELCAGAE KIGSGPCEGD YGGPLICEQH KMRMVLGVIV PGRGCAIPNR PGIFVRVAYY AKWIHKVILT YKL
Content And Storage -20°C
Activity Determined by the dose-dependent stimulation of the proliferation of mouse IMCD3 cells using a concentration range of 10-20 ng/ml.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.