Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse HIN-1/SCGB3A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2954
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2954 100 μg
MAB2954SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2954 Supplier R&D Systems Supplier No. MAB2954
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

HIN-1/SCGB3A1 Monoclonal specifically detects HIN-1/SCGB3A1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen HIN-1/SCGB3A1
Applications Western Blot
Classification Monoclonal
Clone 368203
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q920D7
Gene Alias cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PnSP-2, PNSP2, PnSP-2Uteroglobin-related protein 2, SCGB3A1, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2, UGRP2MGC87867
Gene Symbols Otud4
Host Species Rat
Immunogen E. coli-derived recombinant mouse HIN-1/SCGB3A1 Phe22-Gly104 Accession # Q920D7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92304
Test Specificity Detects mouse HIN-1/SCGB3A1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human HIN-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.