Learn More
Biomatik Corporation Mouse HLA Class II Histocompatibility Antigen, DRB1 Beta Chain (HLA-DRB1) ELISA Kit
Supplier:  Biomatik Corporation EKU05777
Encompass_Preferred
Mouse HLA Class II Histocompatibility Antigen, DRB1 Beta Chain (HLA-DRB1) ELISA Kit for detecting HLA-DRB1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 39.06-2500pg/ml. Sensitivity: 12pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01915. Target Synonyms: HLA-DRB1; MHCDRb1; HLADRb1; Major Histocompatibility Complex Class II DR Beta 1; HLA Class II Histocompatibility Antigen,DRB1-9 Beta Chain; HLA class II histocompatibility antigen, DRB1-15 beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4873
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.