Learn More
Mouse HVEM/TNFRSF14 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | HVEM/TNFRSF14 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2024B |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide |
| Gene Accession No. | NP_849262 |
| Gene Alias | ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.