Learn More
Sino Biological Mouse ICAM1 MAb 25TESTS 50440-R009-P-25
Supplier:  Sino Biological 50440R009P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD54 / ICAM1 (rM CD54 / ICAM1; Catalog#50440-M08H; NP_034623.1; Met 1-Asn 485) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1960
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.