Learn More
FUJIFILM IRVINE SCIENTIFIC INC ACF Mouse IFN-gamma, Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 20016AF100UG
Encompass_Preferred
Interferon gamma (IFN-Y) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-Y is produced by T cells and natural killer cells following antigen-specific activation. IFN-Y binds IFN-Y receptors (IFN-Y R1 and IFN-Y R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-Y-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Mouse IFN-Y is not cross-reactive with human IFN-Y.
Catalog No. 50-130-5789
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.