Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IFN-gamma Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB785SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB785SP 25 μg
MAB785100 100 μg
MAB785500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB785SP Supplier R&D Systems Supplier No. MAB785SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 18 publications

IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IFN-gamma
Applications ELISA
Classification Monoclonal
Clone 37801/37875
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbols IFNG
Host Species Rat
Immunogen E. coli-derived recombinant mouse IFN-gamma
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3458
Test Specificity Detects mouse IFN-gamma in ELISAs. In ELISAs, this antibody does not cross-react with recombinant mouse (rm) C10, rmG-CSF, rmGM-CSF, recombinant human IFN-gamma , rmIL-1 alpha , rmIL-1 beta , rmIL-2, rmIL-3, rmIL-4, rmIL-5, rmIL-6, rmIL-7, rmIL-9, rmIL-10, rmIL-10 R, rmIL-12, rmIL-13, rmJE, rmKC, rmLIF, rmM-CSF, rmMIP-1 alpha , rmMIP-1 beta , rmMIP-2, rmSCF, rmTNF-alpha , rmTpo, and rmVEGF.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.