Learn More
Description
- ELISpot development module provides optimized capture and detection antibody pairings for the quantitative sandwich ELISpot detection of mouse IFN-γ.
- It is an economical alternative to complete kits that allows for assay customization and reduced development time.
- Assay Type: Quantitative sandwich ELISpot development module.
- Species Reactivity: Mouse.
- Assay Capacity: Five 96-well microplates.
- Included Components: Lyophilized anti-mouse IFN-γ capture antibody and lyophilized biotinylated anti-mouse IFN-γ detection antibody.
- Recommended Plates: Nitrocellulose-bottom plates, PVDF-bottom plates, or flat-bottom polystyrene ELISA plates.
- Research Use: For research use only, not for use in diagnostic procedures.
Specifications
Specifications
| Conjugate | Biotin |
| Cross Reactivity | Mouse |
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Form | Lyophilized |
| Product Line | R&D Systems |
| Product Type | ELISpot Development Module |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
