Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Ifng (P01580, 134 amino acids) Partial Recombinant Protein

Catalog No. 89948398 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-398 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-398 Supplier Abnova™ Supplier No. P4450

Used for Func, SDS-PAGE

Specifications

Accession Number P01580
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 15978
Molecular Weight (g/mol) 15.6kDa
Name Ifng (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 100 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with ddH2O, further dilutions should be made into RPMI with 10% FCS, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias IFN-g/IFN-gamma/Ifg
Common Name Ifng
Gene Symbol Ifng
Biological Activity The ED50 as determined by a cytopathic affect inhibition assay with murine NIH 3T3 cells challenged with VSV virus is < 0.1ng/mL, corresponding to a specific activity of > 1 x 107 units/mg.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.